Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022665.3 | NP_073156.3 | 7 - 8 | 904 | 75 | |
XM_008767194.1 | XP_008765416.1 | 8 - 9 | 939 | 75 | ||
GenBank mRNA | X17611.1 | - | 7 - 8 | 903 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |