Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024399.1 | NP_077375.1 | 4 - 5 | 770 | 129 | |
XM_006246815.2 | XP_006246877.1 | 4 - 5 | 812 | 129 | ||
XM_006246817.2 | XP_006246879.1 | 4 - 5 | 812 | 129 | ||
XM_006246818.2 | XP_006246880.1 | 4 - 5 | 812 | 129 | ||
GenBank mRNA | AB023432.1 | - | 4 - 5 | 770 | 129 | |
BC078813.1 | - | 4 - 5 | 772 | 7129010 | 129 | |
BC078814.1 | - | 992 | 7133944 | 129 | ||
FQ232103.1 | - | 4 - 5 | 770 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |