Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236679.2 | XP_006236741.1 | 1 - 2 | 364 | 60 | |
XM_006236684.2 | XP_006236746.1 | 1 - 2 | 364 | 60 | ||
GenBank mRNA | AB018049.1 | - | 1 - 2 | 236 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |