Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031581.2 | NP_113769.1 | 1 | 1213 | 73 | |
GenBank mRNA | U42388.1 | - | 1 | 1215 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |