Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031832.1 | NP_114020.1 | 4 - 5 | 507 | 85 | |
GenBank mRNA | BC089054.1 | - | 4 - 5 | 495 | 7312891 | 85 |
FQ210101.1 | - | 4 - 5 | 517 | 85 | ||
FQ214799.1 | - | 4 - 5 | 789 | 85 | ||
FQ218337.1 | - | 4 - 5 | 507 | 85 | ||
FQ219929.1 | - | 4 - 5 | 507 | 85 | ||
FQ219987.1 | - | 4 - 5 | 507 | 85 | ||
FQ220174.1 | - | 4 - 5 | 506 | 85 | ||
FQ220215.1 | - | 4 - 5 | 509 | 85 | ||
FQ220224.1 | - | 4 - 5 | 501 | 85 | ||
FQ220313.1 | - | 4 - 5 | 507 | 85 | ||
FQ220316.1 | - | 5 - 6 | 737 | 85 | ||
FQ220361.1 | - | 4 - 5 | 666 | 85 | ||
FQ220369.1 | - | 4 - 5 | 507 | 85 | ||
FQ220450.1 | - | 4 - 5 | 507 | 85 | ||
FQ220487.1 | - | 5 - 6 | 707 | 85 | ||
FQ220505.1 | - | 4 - 5 | 507 | 85 | ||
FQ220541.1 | - | 4 - 5 | 507 | 85 | ||
FQ220563.1 | - | 4 - 5 | 507 | 85 | ||
FQ220590.1 | - | 4 - 5 | 509 | 85 | ||
FQ220652.1 | - | 4 - 5 | 507 | 85 | ||
FQ220701.1 | - | 4 - 5 | 506 | 85 | ||
FQ220837.1 | - | 4 - 5 | 506 | 85 | ||
FQ220917.1 | - | 4 - 5 | 507 | 85 | ||
FQ220938.1 | - | 4 - 5 | 507 | 85 | ||
FQ220946.1 | - | 4 - 5 | 507 | 85 | ||
FQ220952.1 | - | 4 - 5 | 507 | 85 | ||
FQ221377.1 | - | 4 - 5 | 507 | 85 | ||
FQ223467.1 | - | 4 - 5 | 509 | 85 | ||
FQ223525.1 | - | 5 - 6 | 299 | 85 | ||
FQ223531.1 | - | 4 - 5 | 507 | 85 | ||
FQ229211.1 | - | 4 - 5 | 85 | |||
FQ229249.1 | - | 4 - 5 | 507 | 85 | ||
FQ229259.1 | - | 4 - 5 | 507 | 85 | ||
FQ229266.1 | - | 4 - 5 | 507 | 85 | ||
FQ229277.1 | - | 4 - 5 | 504 | 85 | ||
FQ229326.1 | - | 4 - 5 | 507 | 85 | ||
FQ229348.1 | - | 4 - 5 | 507 | 85 | ||
FQ229362.1 | - | 4 - 5 | 507 | 85 | ||
FQ229490.1 | - | 4 - 5 | 507 | 85 | ||
FQ229511.1 | - | 4 - 5 | 507 | 85 | ||
FQ229560.1 | - | 4 - 5 | 509 | 85 | ||
FQ229562.1 | - | 4 - 5 | 509 | 85 | ||
FQ229564.1 | - | 4 - 5 | 509 | 85 | ||
FQ229577.1 | - | 4 - 5 | 507 | 85 | ||
FQ229661.1 | - | 4 - 5 | 507 | 85 | ||
FQ229821.1 | - | 4 - 5 | 507 | 85 | ||
FQ229967.1 | - | 4 - 5 | 509 | 85 | ||
FQ229990.1 | - | 4 - 5 | 509 | 85 | ||
FQ229991.1 | - | 4 - 5 | 509 | 85 | ||
FQ229995.1 | - | 4 - 5 | 507 | 85 | ||
FQ230027.1 | - | 4 - 5 | 507 | 85 | ||
FQ230050.1 | - | 4 - 5 | 510 | 85 | ||
FQ230072.1 | - | 4 - 5 | 507 | 85 | ||
FQ230121.1 | - | 4 - 5 | 514 | 85 | ||
FQ230149.1 | - | 4 - 5 | 509 | 85 | ||
J02962.1 | - | 4 - 5 | 507 | 85 | ||
KF639951.1 | - | 3 - 4 | 467 | 85 | ||
KF639952.1 | - | 3 - 4 | 467 | 85 | ||
KF639956.1 | - | 4 - 5 | 446 | 85 | ||
KF639957.1 | - | 4 - 5 | 446 | 85 | ||
M13697.1 | - | 1 - 2 | 95 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |