Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173434.1 | NP_001166905.1 | 4 - 5 | 855 | 100 | |
NM_001173435.1 | NP_001166906.1 | 3 - 4 | 555 | 100 | ||
NM_053301.4 | NP_445753.1 | 5 - 6 | 1119 | 100 | ||
GenBank mRNA | AJ001517.1 | - | 5 - 6 | 1158 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |