Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053530.2 | NP_445982.1 | 1 | 531 | 71 | |
GenBank mRNA | BC166412.1 | - | 1 | 661 | 9033082 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Both primers and probe map within a single exon |