Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053593.2 | NP_446045.1 | 5 - 6 | 818 | 70 | |
GenBank mRNA | BC070956.1 | - | 5 - 6 | 818 | 7104124 | 70 |
FQ219275.1 | - | 5 - 6 | 794 | 70 | ||
FQ234663.1 | - | 4 - 5 | 447 | 70 | ||
L11007.1 | - | 5 - 6 | 750 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |