Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053929.1 | NP_446381.1 | 13 - 14 | 1569 | 73 | |
XM_006228875.2 | XP_006228937.1 | 14 - 15 | 1671 | 73 | ||
XM_006228876.2 | XP_006228938.1 | 14 - 15 | 2051 | 73 | ||
XM_006228877.2 | XP_006228939.1 | 13 - 14 | 1930 | 73 | ||
XM_006228878.2 | XP_006228940.1 | 10 - 11 | 1236 | 73 | ||
XM_008759103.1 | XP_008757325.1 | 13 - 14 | 1955 | 73 | ||
XM_008759104.1 | XP_008757326.1 | 13 - 14 | 1610 | 73 | ||
GenBank mRNA | AB029559.1 | - | 13 - 14 | 1569 | 73 | |
BC081750.1 | - | 12 - 13 | 1985 | 73 | ||
BC098909.1 | - | 13 - 14 | 1559 | 7109606 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |