Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130816.1 | NP_570829.1 | 3 - 4 | 412 | 90 | |
GenBank mRNA | AB079263.1 | - | 3 - 4 | 412 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |