Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133543.3 | NP_598227.3 | 659 | 62 | ||
GenBank mRNA | BC088090.1 | - | 659 | 7368214 | 62 | |
BC098650.1 | - | 721 | 7371059 | 62 | ||
FQ209576.1 | - | 643 | 62 | |||
FQ209710.1 | - | 677 | 62 | |||
U33501.1 | - | 640 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |