Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prokr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138978.1 | NP_620434.1 | 1 - 2 | 569 | 96 | |
XM_006235059.2 | XP_006235121.1 | 3 - 4 | 975 | 99 | ||
GenBank mRNA | AY089975.1 | - | 1 - 2 | 569 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |