Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172009.1 | NP_742006.1 | 3 - 4 | 1728 | 109 | |
GenBank mRNA | BC094521.1 | - | 1823 | 7315968 | 109 | |
S50879.1 | - | 3 - 4 | 1728 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |