Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172091.2 | NP_742088.1 | 3 - 4 | 303 | 80 | |
NM_172092.2 | NP_742089.1 | 3 - 4 | 311 | 80 | ||
NR_073147.1 | 3 - 4 | 311 | 80 | |||
XM_006247876.2 | XP_006247938.1 | 3 - 4 | 417 | 80 | ||
GenBank mRNA | BC081700.1 | - | 3 - 4 | 275 | 7192550 | 80 |
L04796.1 | - | 3 - 4 | 273 | 80 | ||
M96674.1 | - | 3 - 4 | 313 | 80 | ||
X68692.1 | - | 3 - 4 | 273 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |