Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012522.2 | NP_036654.2 | 9 - 10 | 1088 | 75 | |
GenBank mRNA | D01098.1 | - | 9 - 10 | 1089 | 75 | |
M88344.1 | - | 9 - 10 | 1237 | 75 | ||
M88345.1 | - | 6 - 7 | 1501 | 75 | ||
M88346.1 | - | 8 - 9 | 983 | 75 | ||
M88347.1 | - | 8 - 9 | 983 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |