Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012959.1 | NP_037091.1 | 8 - 9 | 1453 | 72 | |
XM_008760512.1 | XP_008758734.1 | 9 - 10 | 1596 | 72 | ||
XM_008760513.1 | XP_008758735.1 | 9 - 10 | 1460 | 72 | ||
XM_008760514.1 | XP_008758736.1 | 7 - 8 | 1476 | 72 | ||
GenBank mRNA | AJ002072.1 | - | 7 - 8 | 1182 | 72 | |
U59486.1 | - | 8 - 9 | 1498 | 72 | ||
U97142.1 | - | 8 - 9 | 1453 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Rattus norvegicus Probe spans exons |