Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021745.1 | NP_068513.1 | 4 - 5 | 741 | 102 | |
XM_008765261.1 | XP_008763483.1 | 3 - 4 | 1098 | 102 | ||
GenBank mRNA | BC083815.1 | - | 3 - 4 | 619 | 7110420 | 102 |
U18374.1 | - | 4 - 5 | 741 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Rattus norvegicus Probe spans exons |