Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024366.1 | NP_077342.1 | 3 - 4 | 291 | 73 | |
XM_006233925.2 | XP_006233987.1 | 3 - 4 | 595 | 73 | ||
XM_008761679.1 | XP_008759901.1 | 2 - 3 | 186 | 73 | ||
GenBank mRNA | L27421.1 | - | 3 - 4 | 231 | 73 | |
X82188.1 | - | 3 - 4 | 291 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |