Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031149.1 | NP_112411.1 | 4 - 5 | 281 | 120 | |
GenBank mRNA | AB000491.1 | - | 4 - 5 | 281 | 120 | |
BC058462.1 | - | 4 - 5 | 277 | 6891005 | 120 | |
D83521.1 | - | 4 - 5 | 299 | 120 | ||
FQ213263.1 | - | 4 - 5 | 298 | 120 | ||
FQ213735.1 | - | 4 - 5 | 273 | 120 | ||
FQ233831.1 | - | 4 - 5 | 284 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |