Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031612.3 | NP_113800.1 | 2 - 3 | 576 | 71 | |
GenBank mRNA | AF179679.1 | - | 2 - 3 | 536 | 71 | |
BC080843.1 | - | 2 - 3 | 562 | 7099981 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |