Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270586.1 | NP_001257515.1 | 12 - 13 | 1670 | 125 | |
NM_001270587.1 | NP_001257516.1 | 11 - 12 | 1676 | 125 | ||
NM_031650.3 | NP_113838.1 | 12 - 13 | 1775 | 125 | ||
GenBank mRNA | AF147740.1 | - | 11 - 12 | 1556 | 125 | |
AF208545.2 | - | 12 - 13 | 1775 | 125 | ||
AF217450.2 | - | 11 - 12 | 1676 | 125 | ||
AJ271682.1 | - | 12 - 13 | 1776 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |