Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031747.1 | NP_113935.1 | 5 - 6 | 560 | 63 | |
GenBank mRNA | BC061809.1 | - | 5 - 6 | 529 | 5598777 | 63 |
D14437.1 | - | 5 - 6 | 560 | 63 | ||
X71071.1 | - | 5 - 6 | 525 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |