Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031811.3 | NP_113999.2 | 2 - 3 | 235 | 98 | |
GenBank mRNA | AF069306.1 | - | 2 - 3 | 221 | 98 | |
BC059126.1 | - | 2 - 3 | 239 | 6889440 | 98 | |
FQ213939.1 | - | 2 - 3 | 247 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |