Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 8 - 9 | 1192 | 60 | |
GenBank mRNA | AF000578.2 | - | 8 - 9 | 1192 | 60 | |
BC087007.1 | - | 8 - 9 | 1129 | 60 | ||
BC161839.1 | - | 8 - 9 | 1156 | 9101355 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |