Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022294.1 | NP_071630.1 | 11 - 12 | 1838 | 84 | |
GenBank mRNA | AF192401.1 | - | 11 - 12 | 1838 | 84 | |
AF192402.1 | - | 11 - 12 | 1838 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |