Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022507.1 | NP_071952.1 | 15 - 16 | 1635 | 87 | |
XM_006239535.2 | XP_006239597.1 | 12 - 13 | 4130 | 87 | ||
XM_006239536.2 | XP_006239598.1 | 12 - 13 | 3336 | 87 | ||
XM_008764337.1 | XP_008762559.1 | 15 - 16 | 1536 | 87 | ||
GenBank mRNA | J04532.1 | - | 15 - 16 | 1635 | 87 | |
M18332.1 | - | 12 - 13 | 1188 | 87 | ||
U27191.1 | - | 12 - 13 | 1400 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |