Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022629.1 | NP_072151.1 | 3 - 4 | 533 | 90 | |
GenBank mRNA | AF160958.1 | - | 3 - 4 | 533 | 90 | |
FQ210746.1 | - | 625 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |