Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023977.2 | NP_076467.2 | 2 - 3 | 469 | 123 | |
GenBank mRNA | AF311954.1 | - | 2 - 3 | 360 | 123 | |
BC092568.1 | - | 2 - 3 | 469 | 7307277 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |