Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031348.2 | NP_112638.2 | 1 - 2 | 182 | 110 | |
XM_008761614.1 | XP_008759836.1 | 1 - 2 | 205 | 110 | ||
GenBank mRNA | AB026057.2 | - | 1 - 2 | 93 | 110 | |
BC088128.1 | - | 1 - 2 | 182 | 7376326 | 110 | |
FQ227647.1 | - | 1 - 2 | 202 | 110 | ||
FQ234790.1 | - | 1 - 2 | 202 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |