Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053309.1 | NP_445761.1 | 3 - 4 | 566 | 77 | |
GenBank mRNA | AB007689.1 | - | 3 - 4 | 566 | 77 | |
AB007690.1 | - | 3 - 4 | 380 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |