Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053404.2 | NP_445856.1 | 7 - 8 | 624 | 67 | |
XM_006254783.2 | XP_006254845.1 | 6 - 7 | 666 | 67 | ||
GenBank mRNA | AF190798.1 | - | 7 - 8 | 685 | 67 | |
AF192493.1 | - | 6 - 7 | 603 | 67 | ||
AF192494.1 | - | 7 - 8 | 624 | 67 | ||
BC100143.1 | - | 6 - 7 | 608 | 7133708 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |