Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aurkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053749.1 | NP_446201.1 | 5 - 6 | 595 | 80 | |
XM_006246569.2 | XP_006246631.1 | 5 - 6 | 613 | 80 | ||
XM_006246570.2 | XP_006246632.1 | 4 - 5 | 500 | 80 | ||
XM_008767757.1 | XP_008765979.1 | 5 - 6 | 510 | 80 | ||
XM_008767758.1 | XP_008765980.1 | 5 - 6 | 456 | 80 | ||
GenBank mRNA | BC097297.1 | - | 5 - 6 | 513 | 7454285 | 80 |
D89731.1 | - | 5 - 6 | 595 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |