Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080892.1 | NP_543168.1 | 8 - 9 | 1108 | 91 | |
GenBank mRNA | AB036799.1 | - | 8 - 9 | 1108 | 91 | |
BC074008.1 | - | 8 - 9 | 1118 | 7100467 | 91 |
Gene Symbol: LOC103689947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761279.1 | XP_008759501.1 | 1108 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |