Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172063.1 | NP_742060.1 | 7 - 8 | 605 | 68 | |
GenBank mRNA | AB017544.1 | - | 7 - 8 | 605 | 68 | |
BC081708.1 | - | 7 - 8 | 594 | 7191447 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |