Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012528.1 | NP_036660.1 | 1 - 2 | 124 | 121 | |
XM_008767770.1 | XP_008765992.1 | 1 - 2 | 162 | 121 | ||
GenBank mRNA | X74833.1 | - | 1 - 2 | 124 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |