Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012647.1 | NP_036779.1 | 3 - 4 | 599 | 108 | |
GenBank mRNA | M22254.1 | - | 3 - 4 | 599 | 108 | |
X03639.1 | - | 3 - 4 | 599 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |