Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013104.3 | NP_037236.1 | 3 - 4 | 560 | 75 | |
GenBank mRNA | BC099742.1 | - | 3 - 4 | 572 | 7937967 | 75 |
FQ220221.1 | - | 3 - 4 | 630 | 75 | ||
FQ220720.1 | - | 3 - 4 | 631 | 75 | ||
FQ220764.1 | - | 3 - 4 | 633 | 75 | ||
FQ230155.1 | - | 3 - 4 | 636 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |