Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013200.1 | NP_037332.1 | 6 - 7 | 744 | 72 | |
XM_006242180.2 | XP_006242242.1 | 6 - 7 | 1200 | 72 | ||
GenBank mRNA | BC085761.1 | - | 6 - 7 | 756 | 7193798 | 72 |
D43623.1 | - | 5 - 6 | 731 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |