Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053962.3 | NP_446414.3 | 8 - 9 | 908 | 57 | |
XM_008769213.1 | XP_008767435.1 | 7 - 8 | 1406 | 57 | ||
GenBank mRNA | BC088110.1 | - | 8 - 9 | 908 | 7323391 | 57 |
J03863.1 | - | 8 - 9 | 1106 | 57 | ||
Y00752.1 | - | 8 - 9 | 925 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |