Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270807.1 | NP_001257736.1 | 5 - 6 | 860 | 77 | |
NM_001270808.1 | NP_001257737.1 | 5 - 6 | 909 | 77 | ||
NM_001270809.1 | NP_001257738.1 | 5 - 6 | 810 | 77 | ||
NM_001270810.1 | NP_001257739.1 | 5 - 6 | 1496 | 77 | ||
NM_001270811.1 | NP_001257740.1 | 5 - 6 | 1846 | 77 | ||
NM_133386.3 | NP_596877.2 | 4 - 5 | 2100 | 77 | ||
XM_006247787.2 | XP_006247849.1 | 5 - 6 | 1180 | 77 | ||
XM_006247788.2 | XP_006247850.1 | 5 - 6 | 691 | 77 | ||
GenBank mRNA | AB049571.1 | - | 5 - 6 | 908 | 77 | |
AB049572.1 | - | 5 - 6 | 860 | 77 | ||
AB049573.1 | - | 5 - 6 | 807 | 77 | ||
AB049574.1 | - | 5 - 6 | 1466 | 77 | ||
AB049575.1 | - | 4 - 5 | 1613 | 77 | ||
BC081738.1 | - | 4 - 5 | 2065 | 7132896 | 77 | |
DQ486894.1 | - | 5 - 6 | 648 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |