Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133396.2 | NP_596887.1 | 4 - 5 | 742 | 65 | |
XM_006238628.2 | XP_006238690.1 | 4 - 5 | 973 | 65 | ||
GenBank mRNA | AB049402.1 | - | 4 - 5 | 743 | 65 | |
BC081737.1 | - | 5 - 6 | 993 | 65 | ||
BC128727.1 | - | 4 - 5 | 748 | 7111312 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |