Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133611.2 | NP_598295.2 | 8 - 9 | 1611 | 60 | |
XM_008765664.1 | XP_008763886.1 | 5 - 6 | 675 | 60 | ||
GenBank mRNA | AJ315643.1 | - | 8 - 9 | 1619 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |