Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153730.2 | NP_714952.1 | 7 - 8 | 1142 | 64 | |
XM_006249843.2 | XP_006249905.1 | 8 - 9 | 1291 | 64 | ||
XM_006249844.2 | XP_006249906.1 | 8 - 9 | 1565 | 64 | ||
XM_006249845.1 | XP_006249907.1 | 8 - 9 | 1271 | 64 | ||
XM_006249846.2 | XP_006249908.1 | 9 - 10 | 1357 | 64 | ||
XM_008769539.1 | XP_008767761.1 | 8 - 9 | 1330 | 64 | ||
XM_008769540.1 | XP_008767762.1 | 9 - 10 | 1185 | 64 | ||
XM_008769541.1 | XP_008767763.1 | 8 - 9 | 1075 | 64 | ||
GenBank mRNA | AJ133126.1 | - | 7 - 8 | 1184 | 64 | |
BC086983.1 | - | 8 - 9 | 1272 | 7133597 | 64 | |
FQ210179.1 | - | 9 - 10 | 1341 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |