Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172317.2 | NP_758528.1 | 3 - 4 | 99 | 72 | |
XM_006228818.2 | XP_006228880.1 | 3 - 4 | 349 | 72 | ||
XM_006228819.2 | XP_006228881.1 | 3 - 4 | 203 | 72 | ||
XM_006228820.2 | XP_006228882.1 | 3 - 4 | 339 | 72 | ||
GenBank mRNA | AW142066.1 | - | 2 - 3 | 104 | 72 | |
CA505583.1 | - | 5 - 6 | 72 | |||
DY471861.1 | - | 3 - 4 | 471 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |