Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173106.1 | NP_775129.1 | 3 - 4 | 127 | 64 | |
GenBank mRNA | AY154894.1 | - | 3 - 4 | 127 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |