Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012610.2 | NP_036742.2 | 4 - 5 | 938 | 65 | |
GenBank mRNA | X05137.1 | - | 4 - 5 | 938 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Probe spans exons |