Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016996.1 | NP_058692.1 | 3 - 4 | 1071 | 91 | |
GenBank mRNA | AY214122.1 | - | 3 - 4 | 1061 | 91 | |
U10354.1 | - | 3 - 4 | 1071 | 91 | ||
U20289.1 | - | 2 - 3 | 697 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |