Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017037.1 | NP_058733.1 | 2 - 3 | 283 | 72 | |
XM_006246584.2 | XP_006246646.1 | 2 - 3 | 281 | 72 | ||
XM_006246585.2 | XP_006246647.1 | 2 - 3 | 290 | 72 | ||
GenBank mRNA | M69139.1 | - | 2 - 3 | 199 | 72 | |
S55427.1 | - | 2 - 3 | 199 | 72 | ||
X62431.1 | - | 2 - 3 | 283 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |