Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017182.2 | NP_058878.1 | 3 - 4 | 466 | 100 | |
XM_006253573.2 | XP_006253635.1 | 3 - 4 | 501 | 100 | ||
XM_006253574.2 | XP_006253636.1 | 3 - 4 | 591 | 100 | ||
XM_006253575.2 | XP_006253637.1 | 3 - 4 | 501 | 100 | ||
XM_006253576.2 | XP_006253638.1 | 3 - 4 | 501 | 100 | ||
XM_006253577.2 | XP_006253639.1 | 3 - 4 | 501 | 100 | ||
GenBank mRNA | BC089093.1 | - | 3 - 4 | 466 | 7126329 | 100 |
M99065.2 | - | 2 - 3 | 316 | 100 | ||
U79139.1 | - | 3 - 4 | 411 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |