Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239476.2 | XP_006239538.1 | 6 - 7 | 1387 | 60 | |
XM_006239479.2 | XP_006239541.1 | 6 - 7 | 1387 | 60 | ||
GenBank mRNA | AF540946.1 | - | 1 - 2 | 216 | 60 | |
AF540947.1 | - | 1 - 2 | 216 | 60 | ||
AF540948.1 | - | 1 - 2 | 216 | 60 | ||
AF540949.1 | - | 1 - 2 | 216 | 60 | ||
U46007.1 | - | 6 - 7 | 1210 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |